Thermo Scientific Fluorescein-5-Maleimide is a high-performance derivative of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes. Fluorescein-5-Maleimide is a sulfhydryl-reactive derivative of fluorescein that labels proteins and other molecules having free thiols (cysteine side chains)
Properties of Fluorescein-5-Maleimide:
• Alternative names: 5-MF, 5-maleimido-fluorescein
• Chemical name: 1H-Pyrrole-2,5-dione, 1-(3',6'-dihydroxy-3-oxospiro (isobenzofuran-1(3H),9'-(9H)xanthen-5-yl)-
• Molecular weight: 427.36±3
• Excitation source: 488 nm spectral line, argon-ion laser
• Excitation wavelength: 494 nm
• Emission wavelength: 518 nm
• Extinction coefficient: > 80,000 M
-1cm
-1• CAS #: 75350-46-8
• Solubility: Soluble in DMF or DMSO
• Reactive groups: Maleimide, reacts with sulfhydryls at pH 6.5 to 7.5
Applications:• Label antibodies for use as immunofluorescent probes
• Label oligonucleotides for hybridization probes
• Detect proteins in gels and on Western blots
Related Products5-IAF (5-iodoacetamido-fluorescein)For Research Use Only. Not for use in diagnostic procedures.