Thermo Scientific™

Restore™ Fluorescent Western Blot Stripping Buffer

Catalog number: 62299
Have Questions?
Thermo Scientific™

Restore™ Fluorescent Western Blot Stripping Buffer

Catalog number: 62299
Have Questions?
Catalog Number
62299
Unit Size
20 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
6229920 mL
-
62300100 mL
-
62300X44 x 100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Restore Fluorescent Western Blot Stripping Buffer is a gentle and highly effective reagent for quickly removing primary and near-infrared (IR) dye-labeled secondary antibodies from western blots.

Features of Restore Fluorescent Western Blot Stripping Buffer:

Fast—strip blots in only 15 minutes at room temperature
Saves time—no need to run new gels and prepare a new blot
Conserve samples—reprobe the same PVDF membrane for multiple targets
Economical—less expensive than other commercially available stripping buffers
Efficient—effectively strips blots the first time

Restore Fluorescent Western Blot Stripping Buffer enables the reuse of PVDF membranes, simplifying the Western blot optimization process and allowing the same blot to be reprobed with different primary antibodies to detect alternative targets. Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane only (Part No. 22860).

Fluorescence Western blotting is a powerful method for detecting multiple targets at once. Restore Fluorescent Western Blot Stripping Buffer allows re-probing of PVDF membranes, saving time and cost. This is ideal when samples are limited and optimization or analysis with different primary antibodies is required.

Traditional stripping methods may adversely alter or remove the sample proteins from the PVDF membrane during the stripping process or may be effective for removing only low-affinity antibodies. In contrast, Restore Fluorescent Western Blot Stripping Buffer Stripping efficiency exceeds 90% while reprobing efficiency matches or exceeds other supplier's formulations. Also, Restore Fluorescent Western Blot Stripping Buffer is conveniently stored at room temperature and easy to use. Simply dilute buffer 1:5 in water and incubate your membrane for 5 to 20 minutes at room temperature.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Western Blot Stripping Buffer
Detection Method
Fluorescent
Form
Liquid
For Use With (Application)
Western Blot
Membrane Compatibility
PVDF
Product Line
Restore™
Quantity
20 mL

Contents & Storage

Upon receipt store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text