Vanquish™ Solvent Monitors
Actual product may vary
Vanquish™ Solvent Monitors
Vanquish™ Solvent Monitors
Vanquish™ Solvent Monitors
Thermo Scientific™
Vanquish™ Solvent Monitors
Avoid errors in solvent management by using a Thermo Scientific™ Vanquish™ Solvent Monitor with your Thermo Scientific™ Vanquish™ HPLC or UHPLC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSolvent Channels
6230.1310-018
6230.1320-014
Catalog number 6230.1310-01
Price (KRW)
-
Solvent Channels:
8
Enable your Thermo Scientific™ Vanquish™ HPLC or UHPLC system to accurately measure the liquid levels in each solvent channel and waste container using the Thermo Scientific™ Vanquish™ Solvent Monitor. The Vanquish Solvent Monitor keeps operators from inadvertently allowing the HPLC system to run dry, preventing lost time and unnecessary solvent consumption. Proactive notification of full waste bottles can also help avoid spillage. The Vanquish Solvent Monitor allows operators to gain benefits in time, productivity, and safety.
View HPLC/UHPLC resources
  • A choice of either 4 or 8 channels of solvents and waste to be monitored, ensuring security for even the most complex setup
  • Full integration into the Thermo Scientific™ Chromeleon™ 7.3 Chromatography Data System (CDS) and the Thermo Scientific™ Vanquish™ User Interface
  • Clear LED indicators allowing quick determination of system status
Specifications
DescriptionVanquish Solvent Monitor 8 channel
Solvent Channels8
TypeSolvent Monitor
Unit SizeEach