Nalgene™ Polypropylene Scissor-Type Forceps
Nalgene™ Polypropylene Scissor-Type Forceps
Nalgene™ Polypropylene Scissor-Type Forceps
Nalgene™ Polypropylene Scissor-Type Forceps
Thermo Scientific™
Nalgene™ Polypropylene Scissor-Type Forceps
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents.
Have Questions?
Catalog NumberTip Style
6320-0010Straight, Serrated
Catalog number 6320-0010
Price (USD)
229.19
Online Exclusive
343.74
Save 114.55 (33%)
Case of 72
Tip Style:
Straight, Serrated
Price (USD)
229.19
Online Exclusive
343.74
Save 114.55 (33%)
Case of 72
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents. Double-action hinges provide extra leverage and long life. These forceps feature a scissor-type handle with a ratchet to lock tips shut and a handy clamp for chromatography columns.
  • Autoclavable
  • Feature jaws with interlocking teeth to grip objects tightly
Specifications
No. per Pack12
Quantity12/Pk.
TypeForceps
MaterialPolypropylene
Product TypeForceps
StyleScissor Type
Tip StyleStraight, Serrated
Unit SizeCase of 72