Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Polycarbonate Face Shield
Thermo Scientific™

Polycarbonate Face Shield

Wear this adjustable polycarbonate face shield over safety glasses. It provides impact protection and features an optically clear 1/16 in. thick polycarbonate window.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLens TintLength (English)Width (English)Unit Size
6355-0001Clear11.6 in.7.8 in.Case of 4
6355-0001PK
also known as 6355-0001
Clear11.6 in.7.8 in.Each
Catalog number 6355-0001
Price (USD)
510.40
Online Exclusive
670.00
Save 159.60 (24%)
Case of 4
-
Add to cart
Lens Tint:
Clear
Length (English):
11.6 in.
Width (English):
7.8 in.
Unit Size:
Case of 4
Price (USD)
510.40
Online Exclusive
670.00
Save 159.60 (24%)
Case of 4
Add to cart
Ask our AI about this Product

Wear the adjustable Thermo Scientific™ Nalgene™ Polycarbonate Faceshield over safety glasses. With an optically clear window made of 1/16 in. (1.5mm) thick polycarbonate, this faceshield provides impact protection as well as protection from UV light in the 200-360nm range. It is useful in germicidal procedures as well as reading of chromatographic/electrophoretic strips and gels. Complies with OSHA 29CFR1910.1030 and ANSI Z87.1-1989 standards for use with bloodborne pathogens.

Features:
  • Wraparound design offers both front and side protection while height provides coverage from the bottom of the neck to the top of the head
  • Features a comfortable and adjustable polyethylene headdress
  • Biohazard/Transparent
Note:UV protection from 200 to 360nm (UV radiation may cause the polycarbonate to develop a slight yellow/brown tint.)
Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 

 

 

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.
Specifications
DescriptionAdjustable Face Shield
Length (English)11.6 in.
No. per Case4/Cs.
Thickness (English)0.060 in.
Thickness (Metric)1.5 mm
Width (English)7.8 in.
Lens TintClear
MaterialPolycarbonate
Product TypeShield Face PC/PE
Unit SizeCase of 4
Lab Safety and Organization
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link