Nalgene™ Tefzel ETFE, Quick Action Spigot
Nalgene™ Tefzel ETFE, Quick Action Spigot
Nalgene™ Tefzel ETFE, Quick Action Spigot
Nalgene™ Tefzel ETFE, Quick Action Spigot
Nalgene™ Tefzel ETFE, Quick Action Spigot
Thermo Scientific™
Nalgene™ Tefzel ETFE, Quick Action Spigot
For Use With: DS2327-series fluorinated carboys
Have Questions?
Catalog NumberDescriptionDiameter (English) Inner Tubing
6432-0010Quick-Action Spigot ETFE fits 1-1/8 in. spigot boss for fluorinated carboys 1-1/8 inch5/8 in.
Catalog number 6432-0010
Price (USD)
208.60
Online Exclusive
298.00
Save 89.40 (30%)
Case of 2
Add to cart
Description:
Quick-Action Spigot ETFE fits 1-1/8 in. spigot boss for fluorinated carboys 1-1/8 inch
Diameter (English) Inner Tubing:
5/8 in.
Price (USD)
208.60
Online Exclusive
298.00
Save 89.40 (30%)
Case of 2
Add to cart
Employ Thermo Scientific™ Nalgene™ Quick-Action ETFE Tefzel™ Spigots as replacement spigots for Nalgene DS2327-series fluorinated carboys. They are chemically resistant to selected solvents and other chemicals. Body and plug are made of Tefzel Ethylene tetrafluoroethylene. Accepts tubing with 5/8 in. I.D.
Features:
  • Autoclavable (unassembled)
  • Tubing adapter not included
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage of compatible solutions, but direct UV exposure should be avoided.
Specifications
DescriptionQuick-Action Spigot ETFE fits 1-1/8 in. spigot boss for fluorinated carboys 1-1/8 inch
Diameter (English) Inner Tubing5/8 in.
Diameter (Metric) Inner Tubing16 mm
For Use With (Equipment)DS2327-series fluorinated carboys
No. per Case2/Cs.
No. per Pack1 Ea.
MaterialETFE
Product TypeSpigot
Unit SizeCase of 2
Share catalog number, name or link