Nalgene™ Stopcocks
Thermo Scientific™
Nalgene™ Stopcocks
Develop in-house fluid transfer systems for accurate filling or dispensing. These stopcocks provide reliable, accurate liquid flow control.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug Bore
6460-00022 mm
6460-0002PK2 mm
6460-00044 mm
6460-0004PK4 mm
Catalog number 6460-0002
Price (USD)
530.00
Case of 6
Plug Bore:
2 mm
Price (USD)
530.00
Case of 6
Permit reliable, accurate liquid flow control in liquid handling systems by using Thermo Scientific™ Nalgene™ Stopcocks. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.
  • Serrated tubulation on each end accepts 1/2 in. through 5/16 in. tubing
  • Polypropylene with Teflon™ TFE plug
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
No. per Case6
No. per Pack1
For Use With (Equipment)In-House Fluid Transfer System
MaterialPolypropylene
Plug Bore2 mm
Plug MaterialPolypropylene
TypeStopcock
Unit SizeCase of 6