Nalgene™ Three-way Stopcock
Nalgene™ Three-way Stopcock
Thermo Scientific™
Nalgene™ Three-way Stopcock
Experience in-house fluid transfer system flexibility with this three-way stopcock, arranged in a T-shape to simply any two — or all three — arms of the stopcock with ease.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug Bore
6470-00022 mm
6470-0002PK2 mm
6470-00044 mm
6470-0004PK4 mm
Catalog number 6470-0002
Price (USD)
387.00
Case of 4
-
Add to cart
Plug Bore:
2 mm
Price (USD)
387.00
Case of 4
Add to cart
Ask our AI about this Product
Experience in-house fluid transfer system flexibility with a Thermo Scientific™ Nalgene™ Three-way Stopcock, arranged in a T-shape to connect any two — or all three — arms of the stopcock with ease.
  • Tubulated outlets are arranged in a T shape and accepts 1/4 in. through 7/16 in. tubing
  • Polypropylene, with non-seizing, leakproof† Teflon™ TFE plug
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
No. per Case4
No. per Pack1
MaterialPolypropylene
Outlet PortTubulated
Plug Bore2 mm
Plug MaterialPolypropylene
TypeStopcock
Unit SizeCase of 4
nalgene-certificate-of-compliance
Certificates of Compliance
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Nalgene and Nunc products.
Start your request  
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link