1x1 image pixel for data collection
eBioscience™ Cell Proliferation Dye eFluor™ 670
Product Image
Invitrogen™

eBioscience™ Cell Proliferation Dye eFluor™ 670

Cell Proliferation Dye eFluor™ 670 is a red fluorescent dye that can be used to monitor individual cell divisions. ThisRead more
Promo ImagePromotion
Promo code:RFLOW25
Get more, spend less on every flow cytometry run
Fuel discovery without breaking your budget. Save up to 33% on flow cytometry essentials—compensation beads, buffers, assays & more. Use code RFLOW25 at checkout. The more you buy, the more you save. Limited time only!Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
65-0840-85Promo Image500 μg
65-0840-90Promo Image4 x 500 μg
Catalog number 65-0840-85
Price (USD)
148.00
Each
In stock
Add to cart
Quantity:
500 μg
Price (USD)
148.00
Each
Add to cart
Cell Proliferation Dye eFluor™ 670 is a red fluorescent dye that can be used to monitor individual cell divisions. This fluorescent dye binds to any cellular protein containing primary amines, and as cells divide, the dye is distributed equally between daughter cells that can be measured as successive halving of the fluorescence intensity of the dye. Up to 6 generations may be visualized. Cell Proliferation Dye eFluor™ 670 can also be used for long term tracking of labeled cells. Analysis using two-parameter plots may provide better resolution of each generation, especially between undivided cells and the first generation. Cells labeled with Cell Proliferation Dye eFluor™ 670 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers, such as the Foxp3 Transcription Factor Staining Buffer Set (cat. 00-5523) or the IC Fixation Buffer (cat. 00-8222) and Permeabilization Buffer (10X) (cat. 00-8333).

Cell Proliferation Dye eFluor™ 670 has a peak excitation of 647 nm and can be excited by the red (633 nm) laser line. It has a peak emission of 670 nm and can be detected with a 660/20 band pass filter (equivanet to APC, Alexa Fluor™ 647, or eFluor™ 660), making it compatible with applications that utilize GFP.

Cell Proliferation Dye eFluor™ 670 has a molecular weight of 792.6 and is supplied as a lyophilized powder. Each vial may be reconstituted to a stock concentration of 5 mM with 126 μL of anhydrous DMSO; once reconstituted it should be protected from light and stored at -20°C with dessicant. It is recommended to use the reconstituted dye within 6 months and to avoid freeze-thawing.

Reported Application
Flow Cytometric Analysis, Microscopy

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
FormLyophilized powder
Quantity500 μg
ColorRed
Emission630/670
For Use With (Application)Proliferation Assay
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineeBioscience
Product TypeDye
Unit SizeEach

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link