eBioscience™ Indo-1 AM Calcium Sensor Dye
Product Image
Invitrogen™

eBioscience™ Indo-1 AM Calcium Sensor Dye

Indo-1 AM Calcium Sensor Dye is a membrane-permeable dye used for determining changes in calcium concentrations in the cell usingRead more
Have Questions?
Catalog NumberQuantity
65-0856-391 mg
Catalog number 65-0856-39
Price (USD)
162.00
Each
-
Add to cart
Quantity:
1 mg
Price (USD)
162.00
Each
Add to cart
Ask our AI about this Product
Indo-1 AM Calcium Sensor Dye is a membrane-permeable dye used for determining changes in calcium concentrations in the cell using fluorescence microscopy, flow cytometry, fluorescence spectroscopy and fluorescence microplate readers. Once Indo-1 enters the cell, esterases cleave the AM group yielding a membrane-impermeable dye. Unbound Indo-1 has a peak emission at 475 nm. Upon binding calcium, the peak emission shifts down to 410 nm.

In flow cytometry this can be measured over time and can be represented as a ratio of the two emission wavelengths. To optimize the ratio between the two emissions, unbound Indo-1 fluorescence should be collected using a filter above 485nm (525 nm is a good option) while bound Indo-1 fluorescence should be collected using a filter below 400 nm. Because the emission profile of Indo-1 is broad, multicolor flow analysis using eFluor ™ 450, eVolve™ 605 or eVolve™ 655 is not possible, however fluorochrome conjugated antibodies utilizing the 488nm or 633 nm are quite compatible with Indo-1.

Molecular Weight: 1009.91
Peak Excitation: 346 nm
Peak Emission: 475 nm

Indo-1 AM should be reconstituted in high-quality, freshly opened DMSO. Recommended concentration: 1 mM.
Once reconstituted, it should be protected from light and stored at -20°C; avoid freeze-thaws.

Purity
>90% by HPLC

Reported Application
Flow Cytometric Analysis, Microscopy, Immunohistochemical Staining

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Quantity1 mg
ColorBlue
Emission346/475
For Use With (Application)Immunohistochemistry (IHC), Flow Cytometry
For Use With (Equipment)Fluorescence Microscope
Product LineeBioscience
Product TypeDye
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link