Informational:Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Dynabeads™ MyOne™ Carboxylic Acid

Catalog number: 65012
Have Questions?
Invitrogen™

Dynabeads™ MyOne™ Carboxylic Acid

Catalog number: 65012
Have Questions?
Catalog Number
65012
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
6501210 mL
-
650112 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Surface carboxylic acid groups allow covalent amide bond formation to proteins/peptides via primary amino- or sulphydryl groups. Rapid binding chemistry, activation through carbodimide is required. Amino-modified oligonucleotides, antibodies or other ligands can easily be coupled to the beads. Low non-specific binding of nucleic acids. The 1 μm Dynabeads™ MyOne™ have a large surface area, high capacity, efficient magnetic pull and a slow sedimentation rate during incubation. The MyOne™ products are tailor-made for use in automated protocols where high throughput is crucial.

Benefits and Features:
• Ideal for use as a solid support in in vitro diagnostic assays.
• Capture of specific DNA/RNA sequences, proteins or any other analyte.
• High magnetic mobility and low sedimentation rate

Concentration:
10 mg/mL

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Protein Purification and Isolation
Product Type
Carboxylic Acid Magnetic Bead
Elution Volume Min
5 μL
Format
Beads in solution
High-throughput Compatibility
High-throughput Compatible
Product Line
Dynabeads™, MyOne™
Purity or Quality Grade
Research Grade
Quantity
10 mL
Sample Type
Any Sample Type
Shipping Condition
Room Temperature
Surface Functionality
Carboxylic Acid
Diameter (Metric)
1 μm

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text