Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Thermo Scientific™

Nalgene™ 500mL LLDPE Safety Bottle Carrier

Secure bottles and reduce chance of bumping, cracking or chipping with this safety bottle carrier.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsUnit Size
6505-0010Bottles (500 mL)EA
6505-0010PK
also known as 6505-0010
Bottles (500 mL)Each
Catalog number 6505-0010
Price (USD)
845.60
Online Exclusive
1,110.00
Save 264.40 (24%)
EA
Add to cart
Holds:
Bottles (500 mL)
Unit Size:
EA
Price (USD)
845.60
Online Exclusive
1,110.00
Save 264.40 (24%)
EA
Add to cart

Secure bottles and reduce chance of bumping, cracking, or chipping. Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier offers greater rigidity and impact resistance. The sturdy, wide handle is molded in one piece with the base allowing a comfortable grip, even when the carrier is full. This linear low density polyethylene carrier holds up to six standard 500mL reagent bottles in any profile.

  • Cover latches securely to base and allow carriers to stack
  • Dimensions: 14L x 10W x 11 in. H (35 x 25 x 28cm)

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications
DescriptionCarrier Safety Bottle LLDPE 500mL
No. per Case6
No. per Pack1
ColorYellow
HoldsBottles (500 mL)
MaterialLLDPE
Positions6
Product TypeBottle Carrier
StackableYes
Unit SizeEA

Share catalog number, name or link