Dynabeads™ MyOne™ Streptavidin T1
Inquire about OEM or Commercial Supply version of this product here.
Dynabeads™ MyOne™ Streptavidin T1
Invitrogen™

Dynabeads™ MyOne™ Streptavidin T1

Dynabeads MyOne Streptavidin T1 beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for use inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
656012 mL
6560210 mL
65604D50 mL
Catalog number 65601
Price (USD)
686.65
Online Exclusive
744.00
Save 57.35 (8%)
Each
Add to cart
Quantity:
2 mL
Price (USD)
686.65
Online Exclusive
744.00
Save 57.35 (8%)
Each
Add to cart

Dynabeads MyOne Streptavidin T1 beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for use in the isolation and handling of biotinylated nucleic acids, antibodies, and other biotinylated ligands. Their small but uniform size presents a high surface area per mg of beads and a correspondingly high capacity for the target ligand, while the high iron content ensures an efficient magnetic pull. These features, along with a slow sedimentation rate during incubation, contribute to a gentle and rapid magnetic separation. Excellent dispersion abilities and the lack of magnetic remanence make Dynabeads streptavidin products ideal for manual and automated protocols, including microfluidic system protocols.

Benefits of Dynabeads MyOne Streptavidin T1 beads include:
Performance—high sensitivity and reproducibility secure consistent results
Efficiency—fast binding kinetics and high binding capacity
Flexibility—flexible protocols with gentle and efficient liquid-phase reaction kinetics
Automation ready—easy automation for high-throughput needs on the Kingfisher Purification System

Consistency and reproducibility
The monolayer of streptavidin leaves most biotin-binding sites available for the binding of biotin and biotinylated ligands. The specific and defined surface allows for efficient capture, separation, and downstream handling. In addition, the low size distribution (<5%) of the monosized beads and the streptavidin monolayer ensure negligible leakage, providing batch consistency and reproducibility of results.

High binding capacity
The size of the molecule and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and molecule and between molecules. With two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization, one mg of Dynabeads MyOne Streptavidin T1 typically binds:
• 950–1,500 pmol free biotin 
• ∼20 μg biotinylated IgG 
• ∼400 pmol biotinylated peptides 
• ∼20 μg biotinylated dsDNA 
• ∼400 pmol biotinylated ss-oligonucleotides

Applications
Thermo Fisher streptavidin-coupled Dynabeads magnetic beads have been used and cited for a wide variety of applications. Although the beads can be used to isolate any biotinylated targets, these hydrophobic beads are particularly well suited for immunoassays, protein purification, cell isolation, phage display, and biopanning.

Learn more about our streptavidin products

Automation ready with the KingFisher Purification System 
The 1-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher Sample Purification Systems  or liquid handlers.

Commercial supply 
Our manufacturing sites are ISO 13485–certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads MyOne Streptavidin T1 beads in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or see our Dynabeads OEM page.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bead TypeMagnetic Polystyrene
Binding Property950–1,500 pmoles free biotin/mg beads
Certifications/ComplianceISO9001 and ISO13485
ColorBrown
Concentration10 mg/mL
DescriptionRecombinant streptavidin covalently coupled to Dynabeads
Diameter (Metric)1 μm
For Use With (Application)Capture of Biotinylated Targets
For Use With (Equipment)KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput CompatibilityYes
Ligand TypeStreptavidin
Product LineDynabeads™, MyOne™
Product TypeStreptavidin Magnetic Beads
Quantity2 mL
Regulatory StatusFor Research Use Only
Sample TypeAny Sample Type
Shelf Life48 months from date of manufacture
Shipping ConditionAmbient Temperature
Size DistributionCV <5%
SpeciesRecombinant
Surface FunctionalityTosylactivated, Hydrophobic
TargetBiotin
FormatBeads in suspension
Isolation TechnologyMagnetic Beads
Unit SizeEach
Contents & Storage
Dynabeads MyOne Streptavidin T1 are supplied in PBS, pH 7.4, with 0.1% BSA and 0.02% NaN3 . Store at 2–8°C.

Frequently asked questions (FAQs)

Can Dynabeads Streptavidin magnetic beads be boiled?

We do not recommend this as streptavidin becomes hydrophobic and aggregates during denaturation.

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Which streptavidin-conjugated Dynabeads magnetic beads are the right beads for my application?

Which product to choose depends on the properties of your sample, the buffers and solutions applied, as well as the downstream application. In general, all Dynabeads Streptavidin beads can be used in applications involving biotinylated ligands; however, some beads may perform better than others in particular applications due to their characteristics. Dynabeads M-280 Streptavidin beads and Dynabeads MyOne Streptavidin T1 beads are commonly used for protein and nucleic acid applications. Dynabeads M-270 Streptavidin beads and MyOne Streptavidin C1 beads are preferred for nucleic acid diagnostics, specifically with samples that have a high concentration of chaotropic salts, immunoassays involving small biotinylated antigens, and in applications that are not compatible with BSA, as these beads are not blocked with BSA. Dynabeads MyOne Streptavidin beads offer increased binding capacity and slower sedimentation rate, making them ideal for automated applications and when larger amounts of a biotinylated compound or its specific target need to be isolated. Please see the selection guide here ( https://www.thermofisher.com/us/en/home/brands/product-brand/dynal/streptavidin-coupled-dynabeads.html?icid=fr-strep-1).

How do I measure the binding of biotinylated nucleic acid on streptavidin beads?

You can assay the supernatant for unbound nucleic acid to determine the amount of nucleic acid bound to the Dynabeads Streptavidin beads. The concentration of nucleic acids can be checked by measuring the OD or by running them on a gel. Alternatively, the nucleic acids can be labeled radioactively and the concentration measured directly on the beads, or fluorescently and measured in the supernatant.

Can Dynabeads Streptavidin beads be used directly in PCR or real-time PCR reactions?

Our Dynabeads Streptavidin magnetic beads can be used directly in PCR or real-time PCR. However, you would have to empirically optimize the amount of beads to be used per volume of reaction.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection