GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
GFM Pro Gas Flowmeter
Thermo Scientific™

GFM Pro Gas Flowmeter

Measure and monitor the flow of all types of gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter.
Have Questions?
Catalog NumberAccuracy
66002-010±2% of Flow or ± 0.05 mL/min., whichever is greater
Catalog number 66002-010
Price (USD)
1,512.17
Each 1
Add to cart
Accuracy:
±2% of Flow or ± 0.05 mL/min., whichever is greater
Price (USD)
1,512.17
Each 1
Add to cart

Measure and monitor the flow of all types of laboratoty gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter. Specifically designed for use with GC systems, this CE-certified flowmeter has a measurement range of 0.5–500mL/min and measures the volumetric flow of any gas with an accuracy of +/- 2% or +/- 0.2mL/min, whichever is greater. Its easy-to-use interface features an over-range warning indicator and automatic shutoff, and its explosion-proof rating allows use with flammable and explosive gases. The compact ergonomic design enables use as a hand-held or benchtop unit and features side grips for added durability.

Additional Product Information
  • Data output via USB port
  • Calibration: traceable to NIST primary standards
  • Uses two AA batteries
  • Recalibration service available
Specifications
DescriptionElectronic Gas Flowmeter
Accuracy±2% of Flow or ± 0.05 mL/min., whichever is greater
Unit SizeEach 1

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link