Slide-A-Lyzer™ Dialysis Cassettes, 20K MWCO
Slide-A-Lyzer™ Dialysis Cassettes, 20K MWCO
Actual product may vary
Thermo Scientific™

Slide-A-Lyzer™ Dialysis Cassettes, 20K MWCO

Use this 20K MWCO semi-permeable membrane to remove salts and contaminants with maximal sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Volume (Metric)
6603030 mL
660050.5 mL
660033 mL
6601212 mL
Catalog number 66030
Price (USD)/ Each
129.65
Online Exclusive
149.00
Save 19.35 (13%)
Add to cart
Sample Volume (Metric):
30 mL
Request bulk or custom format
Price (USD)/ Each
129.65
Online Exclusive
149.00
Save 19.35 (13%)
Add to cart
Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange, and more complete sample recovery.
Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and more complete sample recovery. Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume
Color-coded—purple cassette frame specifies cassettes with the 20,000 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the Viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: none
Sulfur content: 0.04%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 30mL
MembraneCellulose Membrane, Dialysis
Product LineSlide-A-Lyzer™
Purification TargetBuffer Exchange, Protein
Quantity6 Cassettes
FormatCassette
MWCO20 kDa
Sample Volume (Metric)30 mL
Unit SizeEach
Contents & Storage
Store at room temperature

Share catalog number, name or link