Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Dialysis Cassette Kit |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 3.5 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 1 Kit (8 Cassettes) |
Sample Volume (Metric) | 12 mL |
Unit Size | 8 cassettes |
Description | Dialysis Cassettes |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 3.5 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 10 Cassettes |
Sample Volume (Metric) | 0.5 mL |
Unit Size | 10 cassettes |
Description | Dialysis Cassette Kit |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 3.5 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 1 Kit (10 Cassettes) |
Sample Volume (Metric) | 0.5 mL |
Unit Size | 10 cassettes |
Description | Dialysis Cassettes |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 3.5 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 10 Cassettes |
Sample Volume (Metric) | 3 mL |
Unit Size | 10 cassettes |
Description | Dialysis Cassette Kit |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 3.5 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 1 Kit (10 Cassettes) |
Sample Volume (Metric) | 3 mL |
Unit Size | 10 cassettes |
Description | Dialysis Cassettes |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 3.5 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 8 Cassettes |
Sample Volume (Metric) | 12 mL |
Unit Size | 8 cassettes |
Description | Dialysis Cassettes |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 3.5 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 6 Cassettes |
Sample Volume (Metric) | 30 mL |
Unit Size | 6 cassettes |
Catalog Number | Specifications | Unit Size | Description | Sample Volume (Metric) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
66107 | 8 cassettes | Dialysis Cassette Kit | 12 mL | 187.65 Online exclusive 203.00 Save 15.35 (8%) | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
66333 | 10 cassettes | Dialysis Cassettes | 0.5 mL | 115.65 Online exclusive 125.00 Save 9.35 (7%) | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
66335 | 10 cassettes | Dialysis Cassette Kit | 0.5 mL | 143.65 Online exclusive 155.00 Save 11.35 (7%) | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
66330 | 10 cassettes | Dialysis Cassettes | 3 mL | 117.65 Online exclusive 127.00 Save 9.35 (7%) | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
66332 | 10 cassettes | Dialysis Cassette Kit | 3 mL | 167.65 Online exclusive 179.00 Save 11.35 (6%) | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
66110 | 8 cassettes | Dialysis Cassettes | 12 mL | 153.65 Online exclusive 167.00 Save 13.35 (8%) | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
66130 | 6 cassettes | Dialysis Cassettes | 30 mL | 125.65 Online exclusive 136.00 Save 10.35 (8%) | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|