Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Actual product may vary
Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Thermo Scientific™
Freezer and Refrigerator Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
669529.2 cu. ft. UnitsOpen Wire Shelves
6695-129.2 cu. ft. High-performance UnitsOpen Wire Shelves
669623.3, 45.8, 51.1, 78.8 cu. ft. UnitsOpen Wire Shelves
6696-123.3, 45.8, 51.1 cu.ft. UnitsFull Solid Shelves
669729.2 cu. ft. UnitsOpen Wire Shelves
669851.1 cu. ft. UnitsOpen Wire Shelves
71254.7 cu. ft., 11.5 cu. ft. UnitsOpen Wire Shelves
Catalog number 6695
Price (USD)
-
Request A Quote
For Use With (Equipment):
29.2 cu. ft. Units
Type:
Open Wire Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
  • Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)29.2 cu. ft. Units
HoldsGeneral Freezer Items
TypeOpen Wire Shelves
WarrantyWarranty varies by country, please contact us for details
Exterior FinishEpoxy-coated Steel
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link