Thermo Scientific™
UltiMate™ AFC-3000 Low Flow Rate Kit
Catalog number: 6702.0300
Have Questions?
Thermo Scientific™
UltiMate™ AFC-3000 Low Flow Rate Kit
Catalog number: 6702.0300
Have Questions?
Improve the performance of your Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Fraction Collector for preparative applications at low flow rates by using the Thermo Scientific™ Dionex UltiMate AFC-3000 Low Flow Rate Kit. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.
Catalog Number
6702.0300
Unit Size
Each
For Use With (Equipment)
Ultimate AFC-3000 Fraction Collector
Type
Low Flow Rate Kit
Material
PEEK Drop Former
Price (USD)
Full specifications
TypeLow Flow Rate Kit
For Use With (Equipment)Ultimate AFC-3000 Fraction Collector, Ultimate™ AFC-3000 Fraction Collector
MaterialPEEK Drop Former
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeFor Use With (Equipment)TypeMaterialPrice (USD)
6702.0300Full specifications
EachUltimate AFC-3000 Fraction CollectorLow Flow Rate KitPEEK Drop FormerRequest A Quote
TypeLow Flow Rate Kit
For Use With (Equipment)Ultimate AFC-3000 Fraction Collector, Ultimate™ AFC-3000 Fraction Collector
MaterialPEEK Drop Former
Unit SizeEach
Showing 1 of 1
Improve the performance of your Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Fraction Collector for preparative applications at low flow rates by using the Thermo Scientific™ Dionex UltiMate AFC-3000 Low Flow Rate Kit. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.