Thermo Scientific™
UltiMate™ AFC-3000 Collector Normal Phase Kit
Catalog number: 6702.0400
Have Questions?
Thermo Scientific™
UltiMate™ AFC-3000 Collector Normal Phase Kit
Catalog number: 6702.0400
Have Questions?
Adapt your preparative and semi-preparative chromatography for normal phase. The Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Collector Normal Phase kit is ideal for separation and collection of compounds using apolar solvents. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.
Catalog Number
6702.0400
Unit Size
Each
For Use With (Equipment)
Dionex AFC-3000 Automated Fraction Collector
Type
Kit for Normal Phase LC
Material
Stainless Steel Drop Former
Price (USD)
Full specifications
TypeKit for Normal Phase LC
For Use With (Equipment)Dionex AFC-3000 Automated Fraction Collector, Dionex™ AFC-3000 Automated Fraction Collector
MaterialStainless Steel Drop Former
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeFor Use With (Equipment)TypeMaterialPrice (USD)
6702.0400Full specifications
EachDionex AFC-3000 Automated Fraction CollectorKit for Normal Phase LCStainless Steel Drop FormerRequest A Quote
TypeKit for Normal Phase LC
For Use With (Equipment)Dionex AFC-3000 Automated Fraction Collector, Dionex™ AFC-3000 Automated Fraction Collector
MaterialStainless Steel Drop Former
Unit SizeEach
Showing 1 of 1
Adapt your preparative and semi-preparative chromatography for normal phase. The Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Collector Normal Phase kit is ideal for separation and collection of compounds using apolar solvents. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.