Match columns with specific analyses with Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Column ID Cards.
Have Questions?
Catalog number 6710.1505
Price (USD)/ Each
-
Request A Quote
For Use With (Equipment):
TCC-3000 SD/RS
Type:
Column ID Cards
Enhance compliance with improved documentation. Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Column ID Cards make it easy to track which column was used for a specific analysis.
Specifications
TypeColumn ID Cards
For Use With (Equipment)TCC-3000 SD/RS, TCC-3000SD/RS