Search Thermo Fisher Scientific
Catalog Number | Type |
---|---|
6722.0290 | Column Clip kit |
6732.1904 | Column Holder |
Secure your LC column in the Thermo Scientific™ Vanquish™ Column Compartment module using our column holders (for Vanquish Compartments H and C) and column clips (for Vanquish Column Compartment N).