Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Type | Pre-heater |
Max. Pressure | 1300 bar |
Length (Metric) | 530 mm |
Description | Passive Pre-heater, SST, 3 µL, 0.18 × 530 mm |
For Use With (Equipment) | Vanquish Column Compartments |
Material | Stainless Steel |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
6732.0170 | Each | 519.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The passive pre-heater allows for heating and cooling of the eluent to match the column temperature.