Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAMTLLEDWC RGMDVNSQRA LLVWGIPVNC DEAEIEETLQ AAMPQVSYRM LGRMFWREEN AKAALLELTG AVDYAAIPRE MPGKGGVWKV LFKPPTSDAE FLERLHLFLA REGWTVQDVA RVLGFQNPTP TPGPEMPAEM LNYILDNVIQ PLVESIWYKR LTLFSGRDIP GPGEETFDPW LEHTNEVLEE WQVSDVEKRR RLMESLRGPA ADVIRILKSN NPAITTAECL KALEQVFGSV ESSRDAQIKF LNTYQNPGEK LSAYVIRLEP LLQKVVEKGA IDKDNVNQAR LEQVIAGANH SGAIRRQLWL TGAGEGPAPN LFQLLVQIRE EEAKEEEEEA EATLLQLGLE GHF
PNMA1 encodes a protein that is highly restricted to the brain and testis. Anti-PNMA1 reacts mainly with subnuclear elements (including the nucleoli) and to a lesser degree the cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 37 kDa neuronal protein; Neuron- and testis-specific protein 1; Paraneoplastic antigen Ma1; Paraneoplastic antigen Ma1 homolog; paraneoplastic neuronal antigen MA1
Gene Aliases: MA1; PNMA1
UniProt ID: (Human) Q8ND90, (Rat) Q8VHZ4
Entrez Gene ID: (Human) 9240, (Rat) 170636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support