Thermo Scientific™

Slide-A-Lyzer™ MINI Dialysis Device, 7K MWCO

Catalog number: 69562
Have Questions?
Thermo Scientific™

Slide-A-Lyzer™ MINI Dialysis Device, 7K MWCO

Catalog number: 69562
Have Questions?
Slide-A-Lyzer MINI Dialysis Devices, 7K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules larger than 7,000 Daltons in volumes from 10 to 100 μL.
 
Catalog Number
69562
Unit Size
250 devices
Sample Volume (Metric)
0.1 mL
Quantity
250 Devices
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeCup, Microcentrifuge
Product LineSlide-A-Lyzer™
MWCO7.0 kDa
MembraneCellulose Membrane, Dialysis
FormatMINI Dialysis Device
DescriptionMINI Dialysis Device, 0.1 mL
Purification TargetBuffer Exchange, Protein
Sample Volume (Metric)0.1 mL
Quantity250 Devices
Unit Size250 devices
Catalog Number
Unit Size
50 devices
Sample Volume (Metric)
0.1 mL
Quantity
50 Devices
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeCup, Microcentrifuge
Product LineSlide-A-Lyzer™
MWCO7.0 kDa
MembraneCellulose Membrane, Dialysis
FormatMINI Dialysis Device
DescriptionMINI Dialysis Device, 0.1 mL
Purification TargetBuffer Exchange, Protein
Sample Volume (Metric)0.1 mL
Quantity50 Devices
Unit Size50 devices
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeSample Volume (Metric)QuantityPrice (USD)AvailabilityQuantity
69562Full specifications
250 devices0.1 mL250 Devices
-
Product TypeCup, Microcentrifuge
Product LineSlide-A-Lyzer™
MWCO7.0 kDa
MembraneCellulose Membrane, Dialysis
FormatMINI Dialysis Device
DescriptionMINI Dialysis Device, 0.1 mL
Purification TargetBuffer Exchange, Protein
Sample Volume (Metric)0.1 mL
Quantity250 Devices
Unit Size250 devices
69560Full specifications
50 devices0.1 mL50 Devices
-
Product TypeCup, Microcentrifuge
Product LineSlide-A-Lyzer™
MWCO7.0 kDa
MembraneCellulose Membrane, Dialysis
FormatMINI Dialysis Device
DescriptionMINI Dialysis Device, 0.1 mL
Purification TargetBuffer Exchange, Protein
Sample Volume (Metric)0.1 mL
Quantity50 Devices
Unit Size50 devices
Showing 2 of 2
Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

Slide-A-Lyzer MINI Dialysis Devices allow easy sample addition and removal using a standard laboratory pipette and can be used for single or arrays of samples. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with common chemicals and buffers.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker.

Features of Slide-A-Lyzer MINI Dialysis Devices, 7K MWCO:
Flexible format—enables user to dialyze a single sample (tube) or multiple samples (25 array)
Excellent sample recovery—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
One-step protocol—does not require laborious procedures or expensive equipment
100% leak-tested—innovative design does not permit 'wicking' that can occur in home-made devices
Time saving—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

Using the 0.1 mL MINI Device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

For Research Use Only. Not for use in diagnostic procedures.
Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

The 0.1mL MINI Dialysis Devices are designed to be capped (caps included) and inserted into standard 1.5mL microcentrifuge tubes. Alternatively, as many as 25 MINI Devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple inside a large beaker. Using the 0.1mL MINI Device, 100μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes.

As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume, temperature and agitation rates; as well as the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes and the pH, ionic strength, and solvent characteristic of the buffers being used.

Contents & Storage

Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text