Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product Type | Cup, Dialysis Devices, Float Buoy |
Product Line | Slide-A-Lyzer™ |
MWCO | 10 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device Kit |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Kit Contents | Dialysis Devices, 10 devices, Float buoy, 1 float |
Quantity | 1 Kit (10 Devices) |
Unit Size | 10 devices |
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 10 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Quantity | 50 Devices |
Unit Size | 50 devices |
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 10 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Quantity | 250 Devices |
Unit Size | 250 devices |
Product Type | Cup, Dialysis Units, Microcentrifuge Tubes |
Product Line | Slide-A-Lyzer™ |
MWCO | 10 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device Kit |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Kit Contents | Dialysis Units, 10 devices, Microcentrifuge tubes, 10 tubes |
Quantity | 1 Kit (10 Devices) |
Unit Size | 10 devices |
Product Type | Cup, 15 mL Conical |
Product Line | Slide-A-Lyzer™ |
MWCO | 10 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.5 mL |
Quantity | 25 Devices |
Unit Size | 25 devices |
Product Type | Cup, 50 mL Conical |
Product Line | Slide-A-Lyzer™ |
MWCO | 10 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 2 mL |
Quantity | 25 Devices |
Unit Size | 25 devices |
Catalog Number | Specifications | Unit Size | Description | Product Type | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
69576 | 10 devices | MINI Dialysis Device Kit | Cup, Dialysis Devices, Float Buoy | 1 Kit (10 Devices) | 106.65 Online exclusive 113.00 Save 6.35 (6%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69570 | 50 devices | MINI Dialysis Device | Cup, Microcentrifuge | 50 Devices | 232.65 Online exclusive 246.00 Save 13.35 (5%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69572 also known as 000000 | 250 devices | MINI Dialysis Device | Cup, Microcentrifuge | 250 Devices | 714.65 Online exclusive 760.00 Save 45.35 (6%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69574 | 10 devices | MINI Dialysis Device Kit | Cup, Dialysis Units, Microcentrifuge Tubes | 1 Kit (10 Devices) | 93.65 Online exclusive 99.00 Save 5.35 (5%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88401 | 25 devices | MINI Dialysis Device | Cup, 15 mL Conical | 25 Devices | 267.65 Online exclusive 285.00 Save 17.35 (6%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88404 | 25 devices | MINI Dialysis Device | Cup, 50 mL Conical | 25 Devices | 296.65 Online exclusive 314.00 Save 17.35 (6%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The 0.5mL and 2mL MINI Dialysis Devices fit in and are capped by inserting them into the included 15mL and 50mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI Dialysis Devices, low molecular weight contaminant removal, buffer exchange and desalting, can be accomplished within 4 to 8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.