Pierce™ Spin Cups - Cellulose Acetate Filter
Pierce™ Spin Cups - Cellulose Acetate Filter
Thermo Scientific™
Pierce™ Spin Cups - Cellulose Acetate Filter
Thermo Scientific Pierce Cellulose Acetate Filter Spin Cups are convenient tools for manipulating small volumes of affinity supports (20 toRead more
Have Questions?
Catalog NumberQuantity
6970250 cups
Catalog number 69702
Price (USD)
203.00
Each
In stock
Add to cart
Quantity:
50 cups
Price (USD)
203.00
Each
Add to cart
Thermo Scientific Pierce Cellulose Acetate Filter Spin Cups are convenient tools for manipulating small volumes of affinity supports (20 to 400 μL) for protein purification.
Features of Cellulose Acetate Filter Spin Cups:
Smaller pore size: cellulose acetate filters have smaller pore sizes than paper
Column volume: 800μL
Resin volume: 20 to 400μL
Filter type: cellulose acetate filter, 0.45μm pore size
Cap: collection tube cap fits onto inserted spin cup
Simply add the affinity resin and sample to one of the spin cups, then use a microcentrifuge to efficiently wash away contaminants and elute your purified sample without losing any resin in the process. Spin cups allow you to affinity purify more protein in less time.
Applications:
• Affinity purification or affinity chromatography
• Immunodepletion
• Immunoprecipitation (IP)
• Co-immunoprecipitation (co-IP)
Related Products
Pierce™ Spin Columns - Screw Cap
Pierce™ Spin Cups - Paper Filter
Pierce™ Spin Columns - Snap Cap
Pierce™ Micro-Spin Columns
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeEmpty Column, Cellulose Acetate Filter
DescriptionPierce Spin Cups - Cellulose Acetate Filter
FormatMicrocentrifuge, Spin Column
Quantity50 cups
Product LinePierce™
TypeSpin Cup
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link