Thermo Scientific™
SureSTART™ Glass Crimp Top Headspace Vials, Level 3 High Performance Applications
Catalog number: 6PCV6-1F
Have Questions?
Thermo Scientific™
SureSTART™ Glass Crimp Top Headspace Vials, Level 3 High Performance Applications
Catalog number: 6PCV6-1F
Have Questions?
Use Thermo Scientific™ SureSTART™ Glass Crimp Top Headspace Vials, Performance Level 3, for your volatile gas analyses. Available in 6, 10, and 20 mL clear glass formats, these headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyses. This is a Performance Level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. They are Specification Certified to ensure physical dimensions and autosampler compatibility. These vials meet all requirements of US, EU, JPN Pharmacopeia. Use 20 mm crimp caps and septa to seal these headspace vials. Buy in bulk for greater savings.
Catalog Number
6PCV6-1F
Unit Size
Pack of 100
Volume (Metric) Total
6 mL
Height (Metric)
38 mm
Base Style
Flat Bottom
No. per Pack
100/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total6 mL
Closure TypeCrimp Cap
Height (Metric)38 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable5 mL
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Volume (Metric) Total
10 mL
Height (Metric)
46 mm
Base Style
Flat Bottom
No. per Pack
100/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total10 mL
Closure TypeCrimp Cap
Height (Metric)46 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable8 mL
Unit SizePack of 100
Catalog Number
Unit Size
Case of 5000
Volume (Metric) Total
10 mL
Height (Metric)
46 mm
Base Style
Flat Bottom
No. per Pack
5000/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack5000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total10 mL
Closure TypeCrimp Cap
Height (Metric)46 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable8 mL
Unit SizeCase of 5000
Catalog Number
Unit Size
Case of 1000
Volume (Metric) Total
10 mL
Height (Metric)
46 mm
Base Style
Flat Bottom
No. per Pack
1000/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack1000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total10 mL
Closure TypeCrimp Cap
Height (Metric)46 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable8 mL
Unit SizeCase of 1000
Catalog Number
Unit Size
Pack of 100
Volume (Metric) Total
20 mL
Height (Metric)
75 mm
Base Style
Flat Bottom
No. per Pack
100/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizePack of 100
Catalog Number
Unit Size
Case of 5000
Volume (Metric) Total
20 mL
Height (Metric)
75 mm
Base Style
Flat Bottom
No. per Pack
5000/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack5000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 5000
Catalog Number
Unit Size
Case of 1000
Volume (Metric) Total
20 mL
Height (Metric)
75 mm
Base Style
Flat Bottom
No. per Pack
1000/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack1000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 1000
Catalog Number
Unit Size
Pack of 100
Volume (Metric) Total
20 mL
Height (Metric)
75 mm
Base Style
Round Bottom
No. per Pack
100/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleRound Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizePack of 100
Catalog Number
Unit Size
Case of 5000
Volume (Metric) Total
20 mL
Height (Metric)
75 mm
Base Style
Round Bottom
No. per Pack
5000/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack5000/Pk.
Size (Metric) Opening20 mm
Base StyleRound Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 5000
Catalog Number
Unit Size
Case of 1000
Volume (Metric) Total
20 mL
Height (Metric)
75 mm
Base Style
Round Bottom
No. per Pack
1000/Pk.
Price (USD)
Availability
-
Full specifications
Quantity
MaterialClear Glass
No. per Pack1000/Pk.
Size (Metric) Opening20 mm
Base StyleRound Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 1000
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeVolume (Metric) TotalHeight (Metric)Base StyleNo. per PackPrice (USD)AvailabilityQuantity
6PCV6-1FFull specifications
Pack of 1006 mL38 mmFlat Bottom100/Pk.
-
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total6 mL
Closure TypeCrimp Cap
Height (Metric)38 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable5 mL
Unit SizePack of 100
6PCV10-1FFull specifications
Pack of 10010 mL46 mmFlat Bottom100/Pk.
-
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total10 mL
Closure TypeCrimp Cap
Height (Metric)46 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable8 mL
Unit SizePack of 100
6PCV10-1FFFull specifications
Case of 500010 mL46 mmFlat Bottom5000/Pk.
-
MaterialClear Glass
No. per Pack5000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total10 mL
Closure TypeCrimp Cap
Height (Metric)46 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable8 mL
Unit SizeCase of 5000
6PCV10-1FTFull specifications
Case of 100010 mL46 mmFlat Bottom1000/Pk.
-
MaterialClear Glass
No. per Pack1000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total10 mL
Closure TypeCrimp Cap
Height (Metric)46 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable8 mL
Unit SizeCase of 1000
6PCV20-1FFull specifications
Pack of 10020 mL75 mmFlat Bottom100/Pk.
-
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizePack of 100
6PCV20-1FFFull specifications
Case of 500020 mL75 mmFlat Bottom5000/Pk.
-
MaterialClear Glass
No. per Pack5000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 5000
6PCV20-1FTFull specifications
Case of 100020 mL75 mmFlat Bottom1000/Pk.
-
MaterialClear Glass
No. per Pack1000/Pk.
Size (Metric) Opening20 mm
Base StyleFlat Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 1000
6PCV20-1RFull specifications
Pack of 10020 mL75 mmRound Bottom100/Pk.
-
MaterialClear Glass
No. per Pack100/Pk.
Size (Metric) Opening20 mm
Base StyleRound Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizePack of 100
6PCV20-1RFFull specifications
Case of 500020 mL75 mmRound Bottom5000/Pk.
-
MaterialClear Glass
No. per Pack5000/Pk.
Size (Metric) Opening20 mm
Base StyleRound Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 5000
6PCV20-1RTFull specifications
Case of 100020 mL75 mmRound Bottom1000/Pk.
-
MaterialClear Glass
No. per Pack1000/Pk.
Size (Metric) Opening20 mm
Base StyleRound Bottom
Product LineSureSTART Level 3
Volume (Metric) Total20 mL
Closure TypeCrimp Cap
Height (Metric)75 mm
Certifications/ComplianceSpecification Certified
TypeHeadspace Crimp Glass Vial, Level 3
Marking SpotYes
Volume (Metric) Usable18 mL
Unit SizeCase of 1000
Showing 10 of 12Show more
Features
Choose from a round or flat base
  • Round base vials are compatible with most autosamplers and more easily handled by robotic arms that lift the vial from the tray
  • Flat base vials maximize heating efficiency in manual headspace sampling and are required for use in some instrument models
Performance Level 3 Vials – for High Performance Applications
  • Identified by three waves on the vial
  • Convenient write-on patch
  • Suitable for GC and mass spectrometry (MS) applications
  • Recommended for use with Thermo Scientific™ GC Orbitrap™ MS systems and other high-resolution accurate (HRAM) mass instruments
Specification Certified
  • All Thermo Scientific SureSTART vials are dimensionally verified
  • Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances
  • Uniform and defect-free
Lot Traceable
  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request
The SureSTART Performance Levels
SureSTART vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.
• Level 1: Everyday analysis
Chromatography approved
SureSTART Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
• Level 2: High-throughput analysis
When robustness and reproducibility are key
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
• Level 3: High performance
When sensitivity is a must
SureSTART Level 3 products are packed in a clean environment according to our own hygiene concept and regulations to protect against contaminating factors. The vials contain the lowest levels of extractable metals and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes. The septa are conditioned to provide the lowest level of chromatographic extractables. Choose these products for ultimate method performance and high levels of and sample security and integrity.
All SureSTART vials, closures, and septa are certified to meet physical parameter specifications as part of our manufacturing process.