Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Certifications/Compliance | 21 CFR Part 11 |
Description | Execution of NanoDrop One/OneC Security Suite OQ. Verifies function of Security Administration policies and Audit Manager data logging. |
Display | Digital |
Wavelength Range | 190 to 850 nm |
For Use With (Application) | Spectrophotometry |
For Use With (Equipment) | NanoDrop One/OneC Spectrophotometers |
Includes | Requires separate purchase of NanoDrop One/OneC Validated Installation. |
Sample Volume (Metric) | 1 to 2 μL |
Wavelength Accuracy | ±1 nm |
Unit Size | Each |
Certifications/Compliance | All NanoDrop instruments are approved to CE and UL/CSA. |
Compatibility | DYMO LabelWriter 450 printer, Bluetooth keyboard, mouse and barcode reader |
Concentration | dsDNA: Pedestal: 27,500 ng/μL BSA (IgG): Pedestal: 820 (400) mg/mL |
Connections | Three USB-A ports, Ethernet, Bluetooth™ and Wi-Fi (Only available on instruments with Wi-Fi/Bluetooth support) |
Description | NanoDrop One Microvolume UV-Vis Spectrophotometer with Wi-Fi |
Detection Range | dsDNA: Pedestal: 2.0 ng/μL, Cuvette: 0.2 ng/μL, BSA (IgG): Pedestal: 0.06 (0.03) mg/mL, Cuvette: 0.006 (0.003) mg/mL |
Detector Type | 2048-element CMOS Linear Image Sensor |
Display | 7 in. High Definition Color Display, 1280 x 800 pixels |
Dimensions (L x W x H) | 20 x 25.4 x 32.3 cm (8 x 10 x 12.7 in.) |
Languages | English, French, German, Spanish, Polish, Chinese, Japanese, Korean |
Light Source | Xenon Flash |
Spectral Bandwidth | ≤1.8 nm (FWHM at Hg 254 nm) |
Voltage | 12 VDC |
Wavelength Range | 190 to 850 nm |
For Use With (Application) | Nucleic Acid A260, A260/A280, A260/A230 and Labeled Nucleic Acids;, Protein A280 and A205, Protein Pierce 660, Protein Bradford, Protein BCA, Protein Lowry, Labeled Proteins, OD600, Kinetics, UV-Vis, and Custom Methods |
Keypad | Built-in touchscreen, multipoint capacitive touch |
Measurement Time | 8 seconds |
Pathlength | 0.030 to 1.0 mm (auto-ranging) |
Photometric Accuracy | 3% at 0.97 A, 302 nm, (Absorbance expressed at Abs/mm at 25°C) |
Photometric Range | 10 mm equivalent: Pedestal: 0–550 A; Cuvette: 0–1.5 A |
Power Consumption | Operating: 12 to 18 W; Standby: 5 W |
Processor | Android™ |
Repeatability | Typical: 0.002 A (1.0 mm path) or 1%CV, whichever is greater (SD of 10 individual measurements at 0.97 A) |
Sample Temperature | Temperature Control (cuvette only) 37 °C |
Sample Volume (Metric) | Minimum 1μL |
System Requirements | Windows™ 8.1 and 10, 64 bit |
Wavelength Accuracy | ±1 nm |
Weight (English) | 7.9 lb. |
Weight (Metric) | 3.6 kg |
Unit Size | Each |
Certifications/Compliance | All NanoDrop instruments are approved to CE and UL/CSA. |
Compatibility | DYMO LabelWriter 450 printer, Bluetooth keyboard, mouse and barcode reader |
Concentration | dsDNA: Pedestal: 27,500 ng/μL BSA (IgG): Pedestal: 820 (400) mg/mL |
Connections | Three USB-A ports, Ethernet, Bluetooth™ and Wi-Fi (Only available on instruments with Wi-Fi/Bluetooth support) |
Description | NanoDrop OneC Microvolume UV-Vis Spectrophotometer with Wi-Fi |
Detection Range | dsDNA: Pedestal: 2.0 ng/μL, Cuvette: 0.2 ng/μL, BSA (IgG): Pedestal: 0.06 (0.03) mg/mL, Cuvette: 0.006 (0.003) mg/mL |
Detector Type | 2048-element CMOS Linear Image Sensor |
Display | 7 in. High Definition Color Display, 1280 x 800 pixels |
Dimensions (L x W x H) | 20 x 25.4 x 32.3 cm (8 x 10 x 12.7 in.) |
Languages | English, French, German, Spanish, Polish, Chinese, Japanese, Korean |
Light Source | Xenon Flash |
Spectral Bandwidth | ≤1.8 nm (FWHM at Hg 254 nm) |
Voltage | 12 VDC |
Wavelength Range | 190 to 850 nm |
For Use With (Application) | Nucleic Acid A260, A260/A280, A260/A230 and Labeled Nucleic Acids;, Protein A280 and A205, Protein Pierce 660, Protein Bradford, Protein BCA, Protein Lowry, Labeled Proteins, OD600, Kinetics, UV-Vis, and Custom Methods |
Keypad | Built-in touchscreen, multipoint capacitive touch |
Measurement Time | 8 seconds |
Pathlength | 0.030 to 1.0 mm (auto-ranging) |
Photometric Accuracy | 3% at 0.97 A, 302 nm, (Absorbance expressed at Abs/mm at 25°C) |
Photometric Range | 10 mm equivalent: Pedestal: 0–550 A; Cuvette: 0–1.5 A |
Power Consumption | Operating: 12 to 18 W; Standby: 5 W |
Processor | Android™ |
Repeatability | Typical: 0.002 A (1.0 mm path) or 1%CV, whichever is greater (SD of 10 individual measurements at 0.97 A) |
Sample Temperature | Temperature Control (cuvette only) 37 °C |
Sample Volume (Metric) | Minimum 1μL |
System Requirements | Windows™ 8.1 and 10, 64 bit |
Wavelength Accuracy | ±1 nm |
Weight (English) | 7.9 lb. |
Weight (Metric) | 3.6 kg |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
701-058112 | Each | Execution of NanoDrop One/OneC Security Suite OQ. Verifies function of Security Administration policies and Audit Manager data logging. | Request A Quote | - | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ND-ONE-W | Each | NanoDrop One Microvolume UV-Vis Spectrophotometer with Wi-Fi | 13,300.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ND-ONEC-W | Each | NanoDrop OneC Microvolume UV-Vis Spectrophotometer with Wi-Fi | 15,500.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Move one step closer to your research goals with an ergonomic, standalone spectrophotometer with high-resolution, touch screen interface. Powerful auto-range pathlength technology facilitates accurate measurements for concentrated samples with no need for dilutions. This is your innovative alternative to the NanoDrop 2000/2000c Spectrophotometers.
Acclaro Sample Intelligence Technology
Acclaro technology enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting.
NanoDrop One/Nanodrop OneC PC Software with 21 CFR Part 11 Compliance
NanoDrop One/NanoDrop OneC Enhancements
Advanced Connectivity for Data Storage, Printing and Sharing
NanoDrop OneC Highlights
NanoDrop One/NanoDrop OneC Capabilities
* Wi-Fi model not available in certain countries – consult your local NanoDrop distributor
**Patents US6628382 and US6809826
For research use only; not for diagnostic purposes.