Informational:This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

Dynabeads™ EPEC/VTEC O145

Catalog number: 71007
Have Questions?
Applied Biosystems™

Dynabeads™ EPEC/VTEC O145

Catalog number: 71007
Have Questions?
Catalog Number
71007
Unit Size
2 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
710072 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
For the rapid selective concentration of E.coli O145. Method can be performed manually or can be automated by using a BeadRetriever.

This Product Contains: Dynabeads coated with anti-E.coli O145 antibodies

Sensitivity: Sensitivity is very high and enables detection of 1 viable organism in 25g of sample or as few as 100 organisms / ml after pre enrichment
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Antibody Coated Bead
For Use With (Equipment)
Dynabeads™ MPC™, KingFisher mL Food Protection Purification System
Prep Scale
Small Volume Microbial Samples
Product Line
Dynabeads™
Quantity
2 mL
Shipping Condition
Room Temperature

Contents & Storage

Store in refrigerator at 2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text