VersaTREK™ REDOX™ Media
VersaTREK™ REDOX™ Media
Thermo Scientific™
VersaTREK™ REDOX™ Media
Recover organisms from all patient scenarios, including pediatrics, with just two bottles, reducing inventory and costs and enhancing fastidious organism detection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging TypeMedia Type
7103-4480 mL/BottleVersaTREK REDOX 2 Media
7102-4480 mL/BottleVersaTREK REDOX 1 Media
Catalog number 7103-44
Price (USD)
-
Request A Quote
Packaging Type:
80 mL/Bottle
Media Type:
VersaTREK REDOX 2 Media
Just two bottles are all you need to recover organisms from all patient scenarios, reducing inventory and costs, with Thermo Scientific™ VersaTREK™ REDOX™ Media. There's no need for costly specialty media, as REDOX 1 aerobic media and REDOX 2 anaerobic media use dilution and timing to detect microbes and are the only media FDA-cleared for draws as low as 0.1mL, perfect for pediatric patients. Plus, the highly-enriched REDOX media supports a wide range of organisms, enhancing the VersaTREK System's ability to better recover fastidious organisms.
Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.
  • Largest blood-to-broth dilution ratio in the industry (1:9) to dilute out the effects of antimicrobials, serum host factors and other agents
  • Enhanced, enriched media to support the wide range of organisms found in the microbiology laboratory
  • Available in 40 and 80mL sizes and suitable for all patient populations
  • Only system capable of detecting any gas produced or consumed by organisms, allowing detection of a wider range of both common and fastidious organisms
  • Simplified workflow includes easy scan function for bottle entry, intuitive icons to reduce training time, and no daily QC requirement for maximum efficiency
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Manual/AutomatedAutomated
Media TypeVersaTREK REDOX 2 Media
No. per Pack50 Bottles/Box
Packaging Type80 mL/Bottle
Quantity80 mL
Specimen TypeBlood, Sterile Body Fluid
Volume (Metric) Specimen0.1 to 10 mL
Product TypeMicrobiology Media
Target Organism ClassAnaerobes
Test TypeMicrobial Detection
Unit SizeEach
Contents & Storage
Store away from direct sunlight
MBD-0070-Coronavirus Viral Infections-278x123
Sample – Identify – Test
Find your product mix for coronavirus viral transport and associated viral and bacterial infections
#CoronavirusCare
Learn More 
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link