CaptureSelect™ Biotin Anti-IgG-Fc (Human) Conjugate
CaptureSelect™ Biotin Anti-IgG-Fc (Human) Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-IgG-Fc (Human) Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG-Fc (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part (CH3 domain) of all four human IgG subclasses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103262500500 μg
7103262100100 μg
Catalog number 7103262500
Price (USD)/ 500 µg
2,145.00
-
Add to cart
Quantity:
500 μg
Price (USD)/ 500 µg
2,145.00
Add to cart
CaptureSelect™ Biotin Anti-IgG-Fc (Human) Conjugate
Catalog number7103262500
Price (USD)/ 500 µg
2,145.00
-
Add to cart
CaptureSelect Biotin Anti-IgG-Fc (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part (CH3 domain) of all four human IgG subclasses. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-IgG-Fc (Hu) Conjugate allows you to:
  • Detect, quantitate, and characterize all human IgG antibodies (subclasses 1 to 4), recombinant human IgG antibodies, human IgG-derived Fc-fusion proteins including CH2 deleted (or mutated) Fc formats, and total IgG in human plasma and/or serum samples
  • Avoid cross-binding with other isotypes, such as IgA, IgM, IgD, and IgE, free light chains, or IgG antibodies from non-primate species (for example, bovine, horse, and murine)
Applications for CaptureSelect Biotin Anti-IgG-Fc (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product LineCaptureSelect™
Quantity500 μg
TargetIgG (Fc region)
TypeConjugate
Label or DyeBiotin & Other Haptens
Ligand TypeAntibody
Unit Size500 µg
Contents & Storage
Store at -5°C to -30°C.