Thermo Scientific™

CaptureSelect™ Biotin Anti-LC-kappa (Human) Conjugate

Catalog number: 7103272100
Have Questions?
Thermo Scientific™

CaptureSelect™ Biotin Anti-LC-kappa (Human) Conjugate

Catalog number: 7103272100
Have Questions?
Catalog Number
7103272100
Unit Size
100 µg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
7103272100100 µg
-
7103272500500 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

CaptureSelect™ Biotin Anti-LC-kappa (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of human kappa light chains. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect™ Biotin Anti-LC-kappa (Hu) Conjugate allows you to:

  • Detect, quantitate, and characterize all human antibody isotypes and subclasses (for example, IgG1 to 4, IgM, IgA, IgD, and IgE), and Fab fragments thereof, that contain a kappa light chain; as well as recombinant human Fab fragments and/or Fab and Fab2 fragments obtained after papain or pepsin digestion
  • Avoid cross-binding with human antibodies that possess a lambda light chain or IgG from bovine sources such as FCS
  • Screen antibody-antigen interactions—biotin anti-LC-kappa (Hu) forms a stable [low k(diss)] complex with, for example, human IgG antibodies through bivalent binding

Applications for CaptureSelect™ Biotin Anti-LC-kappa (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Biotin Anti-LC-kappa Conjugate
Conjugate
Biotin
Quantity
100 μg
Product Line
CaptureSelect™
Target
Kappa Light Chain

Contents & Storage

Store at -5°C to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text