CaptureSelect™ Biotin Anti-AAV8 Conjugate
CaptureSelect™ Biotin Anti-AAV8 Conjugate
Thermo Scientific™

CaptureSelect™ Biotin Anti-AAV8 Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAV8 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity to adeno-associated virus particles of serotype 8 (AAV8).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103382100100 μg
7103382500500 μg
Catalog number 7103382100
Price (USD)
628.00
Each
-
Add to cart
Quantity:
100 μg
Recurring order eligible. Learn more »
Price (USD)
628.00
Each
Add to cart
Ask our AI about this Product

CaptureSelect Biotin Anti-AAV8 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity to adeno-associated virus particles of serotype 8 (AAV8). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize AAV8 particles.

Applications for CaptureSelect Biotin Anti-AAV8 Conjugate include ELISA, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeAntibody
Product LineCaptureSelect™
Quantity100 μg
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).
Have questions about this product? Ask our AI assisted search.
I am using the 3500 Genetic Analyzer for Sequence Typing & Fragment Analysis. How can I resolve the error report "Failed to save plate. Reason - java.lang.reflect.InvocatiomTargetException: Java heap space"?
Is the Plate Septa, 96 well (Cat. No. 4315933) compatible with 3500/3500xL Genetic Analyzers?
How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?
Is there a way to automatically upload my data into the Thermo Fisher Cloud app directly from the Applied Biosystems 3500 Genetic Analyzer, instead of using Data Collection Software?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection