Thermo Scientific™
para-JEM™ Antibiotic Supplement
Catalog number: 7133-42
Have Questions?
Thermo Scientific™
para-JEM™ Antibiotic Supplement
Catalog number: 7133-42
Have Questions?
Accurately detect M. avium ssp. paratuberculosis in potentially contaminated specimens using the Thermo Scientific™ para-JEM™ Antibiotic Supplement.
Catalog Number
7133-42
Unit Size
Each
Description
para-JEM Antibiotic Supplement
Packaging Type
25 mL Each
Price (USD)
Full specifications
SpeciesMycobacterium avium subsp. paratuberculosis
Descriptionpara-JEM Antibiotic Supplement
Product Typepara-JEM Antibiotic Supplement
Test TypeMicrobial Detection
No. per Pack5
Packaging Type25 mL Each
Quantity25 mL
Specimen TypeVarious
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPackaging TypePrice (USD)
7133-42Full specifications
Eachpara-JEM Antibiotic Supplement25 mL EachRequest A Quote
SpeciesMycobacterium avium subsp. paratuberculosis
Descriptionpara-JEM Antibiotic Supplement
Product Typepara-JEM Antibiotic Supplement
Test TypeMicrobial Detection
No. per Pack5
Packaging Type25 mL Each
Quantity25 mL
Specimen TypeVarious
Unit SizeEach
Showing 1 of 1
Solid medium culture used to be the standard procedure for detecting M. avium ssp. paratuberculosis (MAP), the causative agent of Johne's Disease, in ruminant fecal samples before para-JEM reagents. Due to the bacterium's long generation time, it can take up to 12 weeks of incubation on solid medium before the bacteria are detected. The para-JEM System is a unique, liquid culture method that combines a full line of reagents with the powerful Thermo Scientific™ VersaTREK™ Automated Microbial Detection System for faster, automated detection of MAP.
The para-JEM System includes a number of unique features:
  • Detects faster—able to recover MAP in as little as 2 to 3 weeks, much earlier than conventional methods*
  • Provides increased sensitivity over conventional methods*
  • Provides semi-quantitation for low, medium and high shedders based on time-to-detection*
  • Reduces labor with fewer false positives, false negatives and contaminants*
  • Improves workflow with most tests concluded at six weeks for high, medium and many low shedders*
Utilize para-JEM reagents with the fully automated VersaTREK Microbial Detection System, which automatically incubates and continuously monitors culture bottles for accurate, reliable organism detection as well as enhanced ergonomics and workflow versatility.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
*Cited as a conclusion from an abstract presented at the 43rd Annual AAVLD Meeting - A New Liquid Culture Method, the TREK ESP Culture System II, for the Rapid Detection of Mycobacterium avium subsp. paratuberculosis in Bovine Fecal Samples, S.J.Shin, et. al, College of Veterinary Medicine Diagnostic Laboratory, Cornell University