1x1 image pixel for data collection
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Nalgene™ BioTransport Carrier
Thermo Scientific™

Nalgene™ BioTransport Carrier

Easily view contents with the Thermo Scientific™ Nalgene™ BioTransport Carrier.
Have Questions?
Catalog NumberMaterial
7135-0001PK
also known as 7135-0001
Polycarbonate
Catalog number 7135-0001PK
also known as 7135-0001
Price (USD)
139.06
Online Exclusive
163.60
Save 24.54 (15%)
Each
Add to cart
Material:
Polycarbonate
Price (USD)
139.06
Online Exclusive
163.60
Save 24.54 (15%)
Each
Add to cart

Transport racks containing tubes or sample holders filled with potentially infectious materials using the Thermo Scientific™ Nalgene™ BioTransport Carrier, which offers excellent visibility of contents.

  • Polycarbonate carrier is tough and break resistant
  • Easy to grasp, molded-in handles
  • Silicone gasket seals tightly
  • Clamps securely hold carrier closed and assure leak-resistant seal
  • Stainless-steel handle available separately (7136-0001)
  • For accidental spills inside of carrier, autoclave with clamps closed

To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications
Certifications/ComplianceMeets OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
DescriptionBioTransport Carrier
Dimensions (L x W x H)14.5 x 7.3 x 6.687 in. (36.8 x 18.4 x 17.0cm)
Height (English)6.687 in.
Length (English)14.5 in.
Length (Metric)36.8 cm
No. per Case4
No. per Pack1
Width (English)7.3 in.
AutoclavableAutoclavable
Height (Metric)17.0 cm
MaterialPolycarbonate
Product TypeBioTransport Carrier
Width (Metric)18.4 cm
Unit SizeEach

Share catalog number, name or link