Stopcock Assembly and Cover for Nalgene™ Transparent Polycarbonate Classic Design Desiccator
Catalog number: 715311-0250
Have Questions?
Stopcock Assembly and Cover for Nalgene™ Transparent Polycarbonate Classic Design Desiccator
Catalog number: 715311-0250
Have Questions?
Get the Stopcock Assembly and Cover replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator, Cat #5311-0250. The Nalgene™ Transparent Polycarbonate Classic Design Desiccator maintains an airtight seal without the need of vacuum grease and can be used under full-vacuum and non-vacuum conditions, and is an excellent alternative to glass.
Catalog Number
715311-0250
Unit Size
Each
Description
Cover for port on dessicator, polycarbonate
Price (USD)
Availability
-
Full specifications
Quantity
Product TypeReplacement Part
DescriptionCover for port on dessicator, polycarbonate
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Stopcock Assembly for dessicator
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionStopcock Assembly for dessicator
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
715311-0250Full specifications
EachCover for port on dessicator, polycarbonate
-
Product TypeReplacement Part
DescriptionCover for port on dessicator, polycarbonate
Unit SizeEach
715311-0010Full specifications
EachStopcock Assembly for dessicator
-
DescriptionStopcock Assembly for dessicator
Unit SizeEach
Showing 2 of 2
Description:
  • Replacement stopcock for Nalgene desiccator item number 5311-0250 
  • Replacement cover for Nalgene desiccator item number 5311-0250 
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.