Thermo Scientific™

PCC-54™ Detergent Concentrate

Catalog number: 72289
Have Questions?
Thermo Scientific™

PCC-54™ Detergent Concentrate

Catalog number: 72289
Have Questions?
Catalog Number
72289
Unit Size
4 x 3 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
3 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
722894 x 3 L
-
722883 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific PCC-54 Detergent Concentrate is a general cleaning agent for use on many laboratory surface materials and glassware, including lenses, mirrors, and reflectors.

Applications of PCC-54 Detergent Concentrate include the removal of:

• Coomassie blue dye from test tubes and quartz cuvettes
• Blood or serum residue from glass or plastic beakers and flasks
• Colored dyes from porcelain mortar and pestles
• HIV virus from glassware, tabletops and flooring
• Radioactive isotopes from benchtops and instrument surfaces

The cleaning detergent is mixture of anionic and nonionic surfactants. PCC-54 Detergent is strong enough to remove biological residues from plastic and glass, yet it is fully biodegradable and rinses away thoroughly to leave no residue. Use a 2% (v/v) solution for routine washing applications or soaking in a warm 20% solution to free particularly stubborn dry residues. Mix two parts 2% PCC-54 Detergent solution with one part ethyl alcohol to remove greases and oils.

Complete removal of all impurities and contaminants from glassware is a necessity for most laboratory applications. For many years, caustic alcohol and chromic acid solutions were used for many substances that could not be cleaned from glassware by ordinary detergents. These strongly acidic or strongly basic cleaning products reduce the lifespan of glassware while exposing the user to potential harm. By contrast, Thermo Scientific PCC-54 and RBS 35™ Products are mildly basic, biodegradable, surfactant concentrates containing anionic and nonionic detergents. The surfactants clean by penetrating between contaminating substances and the surfaces to be cleaned. Glass, plastics, quartz, porcelain and ferrous metal apparatus can be cleaned safely and effectively in a 2% solution (v/v) of the concentrates or a 0.2% solution (w/v) of the solids. PCC-54 and RBS™ Cleaners remove both organic and inorganic materials and metal ions from laboratory equipment. The detergents rinse away completely, leaving no residue.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Product Line
PCC-54™
Composition
PCC-PFree, 2%- 20% Solution
Description
PCC-54 Detergent Concentrate
Quantity
4 Pk.
Volume (Metric)
3 L

Contents & Storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text