RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
Gibco™

RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 medium) was originally developed to culture human leukemic cells in suspension andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7240012010 x 500 mL
72400047500 mL
Catalog number 72400120
Price (HKD)
3,692.00
Each
Add to cart
Quantity:
10 x 500 mL
Customize this product
Price (HKD)
3,692.00
Each
Add to cart
Ask our AI about this Product

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This RPMI is modified as follows:
With
• GlutaMAX™
• Phenol Red
• HEPES


The complete formulation is available.

Using RPMI 1640 Medium
RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 medium contains biotin, vitamin B12 and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell TypeLeukemic Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™, GlutaMAX™
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Quantity10 x 500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutaMAX, HEPES, Phenol Red
Without AdditivesNo Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
four-plex-image-COS7-live-cell promo pod

Follow the 5 steps to publication-quality fixed-cell imaging.
Download your free 5-step guide today ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link