RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
Save up to 31% on your order of cell culture products. View promotion details »
Product Image
Gibco™

RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 medium) was originally developed to culture human leukemic cells in suspension andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
724001465 L
7240015310 L
2 Options
Catalog number 72400146
Price (USD)/ 5 L
542.65
Online exclusive
620.00 
Save 77.35 (12%)
-
Add to cart
Quantity:
5 L
Customize this product
Price (USD)/ 5 L
542.65
Online exclusive
620.00 
Save 77.35 (12%)
Add to cart
RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
Catalog number72400146
Price (USD)/ 5 L
542.65
Online exclusive
620.00 
Save 77.35 (12%)
-
Add to cart
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI is modified as follows:
With
• GlutaMAX™
• Phenol Red
• HEPES

The complete formulation is available.

Using RPMI 1640 Medium
RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 medium contains biotin, vitamin B12 and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

cGMP manufacturing and quality system
RPMI 1640 is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical RPMI 1640 product made in our Scotland facility (72400-054). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell TypeLeukemic Cells
ClassificationAnimal Origin-free
Concentration1 X
FormLiquid
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Serum LevelStandard Serum Supplementation
Shelf Life12 Months From Date of Manufacture
SterilitySterile-filtered
With AdditivesGlutaMAX, HEPES, Phenol Red
Without AdditivesNo Sodium Pyruvate
Product LineGibco™, GlutaMAX™, SynerGy™
Quantity5 L
Shipping ConditionRoom Temperature
Unit Size5 L
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture