Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 15 mL |
Particle Density | 1.05g/cm³ |
Preservative | 0.05% Na Az |
Quantity | 15 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 3 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 15 mL |
Particle Density | 1.05g/cm³ |
Preservative | 0.05% Na Az |
Quantity | 15 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 2 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 100 mL |
Particle Density | 1.05g/cm³ |
Preservative | 0.05% Na Az |
Quantity | 100 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 2 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 1000 mL |
Particle Density | 1.05g/cm³ |
Preservative | 0.05% Na Az |
Quantity | 1000 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 2 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 100 mL |
Particle Density | 1.05g/cm³ |
Preservative | 0.05% Na Az |
Quantity | 100 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 3 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
Color | White or Colorless |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 15 mL |
Particle Density | 1.05g/cm³ |
Preservative | Pasteurized |
Quantity | 15 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 0.2 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
Color | White or Colorless |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 100 mL |
Particle Density | 1.05g/cm³ |
Preservative | Pasteurized |
Quantity | 100 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 0.2 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
Color | White or Colorless |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 1000 mL |
Particle Density | 1.05g/cm³ |
Preservative | Pasteurized |
Quantity | 1000 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | High Acid |
Diameter (Metric) | 0.2 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
Color | White or Colorless |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 15 mL |
Particle Density | 1.05g/cm³ |
Preservative | Pasteurized |
Quantity | 15 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | Medium Acid |
Diameter (Metric) | 0.2 μm |
Unit Size | Each |
Additive | Proprietary anionic surfactant |
Certifications/Compliance | Includes Certificate of Analysis |
Concentration | 10% |
Description | Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications. |
Color | White or Colorless |
For Use With (Application) | Covalent Coupling |
Material | Polystyrene Sulfate |
Product Type | Specialty Particles |
Includes | Package Insert Sheet containing Certicate of Analysis |
Packaging | 100 mL |
Particle Density | 1.05g/cm³ |
Preservative | Pasteurized |
Quantity | 100 mL |
Sample Evaluation Packs | Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles. |
Specialty Packaging | Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request. |
Surface Acid Loading | Medium Acid |
Diameter (Metric) | 0.2 μm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Diameter (Metric) | Surface Acid Loading | Concentration | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
73003420100250 also known as MCG-73003420100250-EACH | Each | 15 mL | 3 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
73003305100250 | Each | 15 mL | 2 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
73003305100350 | Each | 100 mL | 2 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
73003305100450 | Each | 1000 mL | 2 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
73003420100350 | Each | 100 mL | 3 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
83000520100290 also known as MCG-83000520100290-EACH | Each | 15 mL | 0.2 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
83000520100390 | Each | 100 mL | 0.2 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
83000520100490 | Each | 1000 mL | 0.2 μm | High Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
83000550100290 | Each | 15 mL | 0.2 μm | Medium Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
83000550100390 | Each | 100 mL | 0.2 μm | Medium Acid | 10% | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Parking Area Properties
Recommended for:
Note:
When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.