GeneTitan™ ZeroStat AntiStatic Gun
Product Image
Applied Biosystems™
GeneTitan™ ZeroStat AntiStatic Gun
The GeneTitan™ ZeroStat Anti-Static Gun is designed for use with the GeneTitan Multi-Channel (MC) Instrument.GeneTitan Multi-Channel (MC) Instrument automates arrayRead more
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Catalog NumberQuantity
74-0014Promo Image1 each
Catalog number 74-0014
Price (USD)
215.00
Each
Add to cart
Quantity:
1 each
Price (USD)
215.00
Each
Add to cart
The GeneTitan™ ZeroStat Anti-Static Gun is designed for use with the GeneTitan Multi-Channel (MC) Instrument.
GeneTitan Multi-Channel (MC) Instrument automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and state-of-the art imaging device into a single bench-top instrument.
The imaging device in GeneTitan MC Instrument uses an external, high-intensity xenon lamp and dual excitation and emission filters to capture images from array plates for use in genome-wide expression and genotyping studies.
GeneTitan MC Instrument provides the highest throughput and laboratory productivity per technician by minimizing user intervention and allowing unattended, overnight processing of large numbers of samples in parallel. This unique degree of hands-free automation means your data will be consistently reproducible, your lab will be more productive, and you can spend less time acquiring and managing data and have more time for science.
Key benefits
Easy – Designed for operators with minimal microarray experience
Flexible – Supports gene expression on 16-, 24-, 96-, array layouts and genotyping studies on 96- and 384-array layouts
Accurate – Delivers high-quality, consistent data every time
Efficient – Condenses hands-;on processing time to as little as 30 minutes, images an array in less than 5 minutes, and operates unattended overnight
Robust – Has fewer moving parts so it's easily maintained
Scalable – Meets both medium- and high-throughput needs
Adaptable – Creates flexible workflows and sample registration via Affymetrix GeneChip Command Console Software (AGCC).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)GeneTitan Multi-Channel (MC) Instrument
Quantity1 each
Voltage240 V
TypeImaging Device
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link