1x1 image pixel for data collection
Microlite™ White Microtiter™ Plates, Plate, Microlite 2
Microlite™ White Microtiter™ Plates, Plate, Microlite 2
Microlite™ White Microtiter™ Plates, Plate, Microlite 2
Microlite™ White Microtiter™ Plates, Plate, Microlite 2
Microlite™ White Microtiter™ Plates, Plate, Microlite 2
Thermo Scientific™
Microlite™ White Microtiter™ Plates, Plate, Microlite 2
Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Microlite Microtiter Plates.
Have Questions?
Catalog number 7417
Price (USD)
-
Request A Quote
Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Thermo Scientific™ Microlite White Microtiter Plates.
Available in plates or 12-well breakable strips.
  • Polystyrene
  • Choice of two surfaces, each performance assayed using an IgG binding assay
SURFACES
  • Microlite 1+ - Hydrophobic surface for binding of biomolecules with hydrophilic domains
  • Microlite 2+ - Slightly hydrophilic surface for increased binding of biomolecules with hydrophilic/hydrophobic character such as many proteins
Note:
Other configurations are available in Microlite 1 and 2 formulations.
Specifications
FormatStandard
ColorWhite
MaterialPolystyrene
Plate BlockingNone
Detection MethodChemiluminescent
Target MoleculeAntibodies (Glycoproteins) and Proteins
SterilityNon-sterile
Binding PropertySlightly Hydrophilic
No. of Wells96
Array8 x 12
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link