Informational:Regulatory requirements vary by country; products may not be available in your geographic area.
Applied Biosystems™

PrioCHECK™ L. hardjo Ab Strip Kit

Catalog number: 7442080
Have Questions?
Applied Biosystems™

PrioCHECK™ L. hardjo Ab Strip Kit

Catalog number: 7442080
Have Questions?
Catalog Number
7442080
Unit Size
440 samples
Price (USD)
Catalog NumberUnit SizePrice (USD)
7442080440 samples
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Note: The name of this product has changed. It was previously called PRIOCHECK LEPTOSPIRA HARDJO AB ELISA.

The PrioCHECK™ L. hardjo Antibody ELISA is an indirect ELISA for the detection of anti-L. hardjo antibodies. The test is suited for large-scale screening of serum and (bulk) milk samples. Test samples are incubated in plates coated with the inactivated L. hardjo antigen. Plates are then washed and an anti-bovine antibody labeled with an enzyme that generates a color signal is added. The signal is measured and if color develops the sample is positive for anti-L. hardjo antibodies. Color intensity is directly related to the amount of anti-L. hardjo antibodies present in the test sample.

Specifications

Assay Sensitivity
See product documentation
Immunoassay Kit Format
Other Format
Product Line
PrioCHECK™, Prionics™
Product Type
Strip Kit
Protein Family
Other Proteins
Sample Type
Serum
Target Species
Bovine
Gene ID (Entrez)
Non-Gene
Gene Symbol
Non-Gene
Immunoassay Kit Grade
Strip
Quantity
5 Plates
Target Kit Named
Leptospira Hardjo Antibody
Target Organism Class
Leptospira interrogans serovar hardjo (L. hardjo)
Target Species Validated
Bovine

Contents & Storage

5 Plates

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text