Search Thermo Fisher Scientific
Perform bioprocessing or separation of large volumes with very low solids content using the Thermo Scientific™ TCF-20 Continuous Flow Rotor. Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to Thermo Scientific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid—shorten run set-up time while providing peace-of-mind the rotor is secure.
Auto-Lock Rotor Exchange
Auto-ID Instant Rotor Identification
Speed Handle