Benchtop Rotors
Benchtop rotors are designed for use in benchtop centrifuges. Rotors are available in a wide variety of models, and for a wide range of tube and bucket capacities. Choose from a fixed-angle, swing-out, drum, and horizontal rotors; some models have lids. The Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor facilitates high-volume microplate processing