2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
Thermo Scientific™

2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge

Thermo Scientific™ 2000 mL Bottle designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityMaterial
75003870Non-sterilePPCO
75003872Non-sterilePolypropylene
75003871SterilePolypropylene
Catalog number 75003870
Price (USD)
640.37
Online Exclusive
738.00
Save 97.63 (13%)
Pack of 2
-
Add to cart
Sterility:
Non-sterile
Material:
PPCO
Price (USD)
640.37
Online Exclusive
738.00
Save 97.63 (13%)
Pack of 2
Add to cart
Ask our AI about this Product
Process a 16L harvest in a single run with the Thermo Scientific™ 2000 mL Bottle, the ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
The Thermo Scientific™ 2000 mL Bottles are ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge.
Specifications
Description2000 mL oval bottle
For Use With (Application)For mammalian, yeast and bacterial cell separations
For Use With (Equipment)Sorvall BIOS 16 Bioprocessing Centrifuge
No. per CaseCase of 2
TransparencyNo
Capacity (Metric)2000 mL
Certifications/ComplianceFood Grade
ColorWhite
MaterialPPCO
SterilityNon-sterile
Unit SizePack of 2
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
5397-2827Certificate of AnalysisSep 20, 202275003872
5344-2773Certificate of AnalysisSep 20, 202275003872
2773Certificate of AnalysisJun 20, 202275003872
5063-2580Certificate of AnalysisJun 01, 202275003872
2580Certificate of AnalysisMay 31, 202275003872
5 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

Share catalog number, name or link

1x1 image pixel for data collection