2000mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
2000mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
Actual product may vary
Thermo Scientific™
2000mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
Thermo Scientific™ 2000 mL Bottle designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
Have Questions?
Change viewbuttonViewtableView
Sterility:
Non-sterile
Sterilized using Beta irradiation following standards ISO 11137-1 and ISO 11137-2 (non-sterile)
Catalog number 75003872
Price (USD)/ Case of 2
-
Request A Quote
Sterility:
Non-sterile
Process a 16L harvest in a single run with the Thermo Scientific™ 2000 mL Bottle, the ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
The Thermo Scientific™ 2000 mL Bottles are ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge.
Specifications
Capacity2000 mL
Capacity (Metric)2000 mL
Capacity (English)67.62 oz.
ColorWhite
Description2000 mL bottle
For Use With (Application)For mammalian, yeast and bacterial separations
MaterialPolypropylene
QuantitySet of 2
SterilityNon-sterile
TransparencyTranslucent
Certifications/ComplianceUSP Class VI Non-Cytotoxic
AutoclavableAutoclavable
GraduatedYes
Max. Force7187 x g
MouthWide
ShapeConical
Product TypeBioprocess Container
For Use With (Equipment)BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swinging Bucket Rotor
Closure MaterialPolypropylene
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCase of 2