Thermo Scientific™

Beaker Racks for Solaris™ Open Air Shakers & Temperature Controlled Shakers, and MaxQ™ 6000 Shakers

Catalog number: 75004118
Have Questions?
Thermo Scientific™

Beaker Racks for Solaris™ Open Air Shakers & Temperature Controlled Shakers, and MaxQ™ 6000 Shakers

Catalog number: 75004118
Have Questions?

Mix and match Thermo Scientific™ Solaris shaker accessories to suit individual needs in the laboratory.  Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers and MaxQ6000 Shakers.

 
Catalog Number
75004118
Unit Size
Each
Description
Rack for 100 mL beaker
Dimensions (L x W x H)
355 x 288 x 40 mm
Holds
16 Beakers
Price (USD)
Full specifications
Holds16 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)355 x 288 x 40 mm
DescriptionRack for 100 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 30 mL beaker
Dimensions (L x W x H)
400 x 210 x 25 mm
Holds
32 Beakers
Price (USD)
Full specifications
Holds32 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 25 mm
DescriptionRack for 30 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 100 mL beaker
Dimensions (L x W x H)
400 x 210 x 40 mm
Holds
15 Beakers
Price (USD)
Full specifications
Holds15 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 40 mm
DescriptionRack for 100 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 250 mL beakers, 9 x 18 in. platform
Dimensions (L x W x H)
400 x 210 x 58 mm
Holds
8 Beakers
Price (USD)
Full specifications
Holds8 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 58 mm
DescriptionRack for 250 mL beakers, 9 x 18 in. platform
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 400 mL beaker
Dimensions (L x W x H)
400 x 210 x 58 mm
Holds
6 Beakers
Price (USD)
Full specifications
Holds6 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 58 mm
DescriptionRack for 400 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 600 mL beaker
Dimensions (L x W x H)
400 x 210 x 64 mm
Holds
4 Beakers
Price (USD)
Full specifications
Holds4 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 64 mm
DescriptionRack for 600 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 30 mL beaker
Dimensions (L x W x H)
350 x 286 x 25 mm
Holds
36 Beakers
Price (USD)
Full specifications
Holds36 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)350 x 286 x 25 mm
DescriptionRack for 30 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 250 mL beaker
Dimensions (L x W x H)
351 x 286 x 58 mm
Holds
9 Beakers
Price (USD)
Full specifications
Holds9 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)351 x 286 x 58 mm
DescriptionRack for 250 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 400 mL beaker
Dimensions (L x W x H)
351 x 286 x 58 mm
Holds
8 Beakers
Price (USD)
Full specifications
Holds8 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)351 x 286 x 58 mm
DescriptionRack for 400 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Rack for 600 mL beaker
Dimensions (L x W x H)
351 x 286 x 64 mm
Holds
5 Beakers
Price (USD)
Full specifications
Holds5 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)351 x 286 x 64 mm
DescriptionRack for 600 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionDimensions (L x W x H)HoldsPrice (USD)
75004118Full specifications
EachRack for 100 mL beaker355 x 288 x 40 mm16 BeakersRequest A Quote
Holds16 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)355 x 288 x 40 mm
DescriptionRack for 100 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004110Full specifications
EachRack for 30 mL beaker400 x 210 x 25 mm32 BeakersRequest A Quote
Holds32 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 25 mm
DescriptionRack for 30 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004112Full specifications
EachRack for 100 mL beaker400 x 210 x 40 mm15 BeakersRequest A Quote
Holds15 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 40 mm
DescriptionRack for 100 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004113Full specifications
EachRack for 250 mL beakers, 9 x 18 in. platform400 x 210 x 58 mm8 BeakersRequest A Quote
Holds8 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 58 mm
DescriptionRack for 250 mL beakers, 9 x 18 in. platform
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004114Full specifications
EachRack for 400 mL beaker400 x 210 x 58 mm6 BeakersRequest A Quote
Holds6 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 58 mm
DescriptionRack for 400 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers
Unit SizeEach
75004115Full specifications
EachRack for 600 mL beaker400 x 210 x 64 mm4 BeakersRequest A Quote
Holds4 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform9 x 18 in.
Dimensions (L x W x H)400 x 210 x 64 mm
DescriptionRack for 600 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004116Full specifications
EachRack for 30 mL beaker350 x 286 x 25 mm36 BeakersRequest A Quote
Holds36 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)350 x 286 x 25 mm
DescriptionRack for 30 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004119Full specifications
EachRack for 250 mL beaker351 x 286 x 58 mm9 BeakersRequest A Quote
Holds9 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)351 x 286 x 58 mm
DescriptionRack for 250 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004120Full specifications
EachRack for 400 mL beaker351 x 286 x 58 mm8 BeakersRequest A Quote
Holds8 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)351 x 286 x 58 mm
DescriptionRack for 400 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
75004121Full specifications
EachRack for 600 mL beaker351 x 286 x 64 mm5 BeakersRequest A Quote
Holds5 Beakers
Product TypeBeaker Rack
Dimensions (LxW) Platform11 x 14 in. or 12 x 14 in.
Dimensions (L x W x H)351 x 286 x 64 mm
DescriptionRack for 600 mL beaker
For Use With (Equipment)Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
Unit SizeEach
Showing 10 of 12Show more

Mix and match Thermo Scientific™ Solaris shaker accessories to suit individual needs in the laboratory

  • Racks can hold beakers from 30 - 600 mL
  • Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers and MaxQ6000 Shakers
  •  

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text