Adapters for TX-1000 Swinging Bucket Rotor
Adapters for TX-1000 Swinging Bucket Rotor
Thermo Scientific™
Adapters for TX-1000 Swinging Bucket Rotor
An assortment of adapters for outstanding flexibility
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityFor Use With (Equipment)
750053924 x 250 mL250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
750073034 x 6 mL Standard or 4 x 2 mL Deep-WellMicroplate carriers
7500730696 x 15 mL15mL Conical Tube
Catalog number 75005392
Price (USD)
1,124.63
Online Exclusive
1,280.00
Save 155.37 (12%)
Pack of 4
In stock
Add to cart
Capacity:
4 x 250 mL
For Use With (Equipment):
250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
Recurring order eligible. Learn more »
Price (USD)
1,124.63
Online Exclusive
1,280.00
Save 155.37 (12%)
Pack of 4
Add to cart
Ask our AI about this Product
An assortment of adapters for outstanding flexibility
Specifications
For Use With (Equipment)250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
Max. Tube Diameter62 mm
Max. Tube Length145 mm
Tube Type250 mL Conical Bottle
Capacity4 x 250 mL
TypeRotor Adapter
Unit SizePack of 4
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection