You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Actual product may vary
Thermo Scientific™
Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Have Questions?
Change view
Catalog Number
Description
75005723
Unsealed Buckets (set of 2)
75005721
Sealed Bucket; Capacity: 4 Standard or 2 Midi-Deepwell plates (Set of 2)
2 Options
Catalog number
75005723
Price (USD)
/ Pack of 2
1,107.00
Online exclusive
1230.00
Save 123.00 (10%)
-
Add to cart
Description:
Unsealed Buckets (set of 2)
Price (USD)
/ Pack of 2
1,107.00
Online exclusive
1230.00
Save 123.00 (10%)
Add to cart
Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Catalog number
75005723
Price (USD)
/ Pack of 2
1,107.00
Online exclusive
1230.00
Save 123.00 (10%)
-
Add to cart
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Max Tube Dimensions: Height <33 mm
Specifications
For Use With (Equipment)
Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Capacity
4 Standard or 2 Midi-Deepwell plates
Description
Unsealed Buckets (set of 2)
Unit Size
Pack of 2